7-Fluoroindoline 95%

Lieferant: Apollo Scientific

PC540097-250MG
APOSPC540097-250MGEA 144 EUR
APOSPC540097-250MG
7-Fluoroindoline 95%
7-Fluoroindoline
Formel: C₈H₈FN·ClH
Molecular Weight: 173,615 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09041825
CAS-Nummer: 769966-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR