4-Brom-2-fluorbenzolsulfonamid

Lieferant: Apollo Scientific

PC5417-1G
APOSPC5417-1GEA 23.2 EUR
APOSPC5417-1G
4-Brom-2-fluorbenzolsulfonamid
4-Brom-2-fluorbenzolsulfonamid
Formel: C₆H₅BrFNO₂S
MDL: MFCD03094291
CAS-Nummer: 214210-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR