2,6-Dimethyl-4-fluorobenzyl alcohol

Lieferant: Apollo Scientific

PC5427-1G
APOSPC5427-1GEA 452 EUR
APOSPC5427-1G
2,6-Dimethyl-4-fluorobenzyl alcohol
2,6-Dimethyl-4-fluorobenzyl alcohol
Formel: C₉H₁₁FO
Molecular Weight: 154.181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701056
CAS-Nummer: 773868-67-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR