Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane

Lieferant: Apollo Scientific
Achtung

PC5428-10G PC5428-5G
APOSPC5428-10GEA 34.8 EUR
APOSPC5428-10G APOSPC5428-5G
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Formel: C₂₇H₁₆F₆N₂O₆
Molecular Weight: 578.43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00317556
CAS-Nummer: 69563-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR