Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Lieferant: Apollo Scientific
|
Achtung
|
PC5428-10G
PC5428-5G
APOSPC5428-10GEA
34.8
EUR
APOSPC5428-10G
APOSPC5428-5G
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Formel:
C₂₇H₁₆F₆N₂O₆ Molecular Weight: 578.43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00317556 CAS-Nummer: 69563-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...