2-(Trifluoromethyl)-DL-phenylglycine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-DL-Phg(2-CF₃)-OH, 2-[Amino(carboxy)methyl]benzotrifluoride, 2-Amino-2-(2-(trifluoromethyl)phenyl)acetic acid, Amino[2-(trifluoromethyl)phenyl]acetic acid

PC5438-1G PC5438-500MG PC5438-5G PC5438-10G
APOSPC5438-1GEA 21.9 EUR
APOSPC5438-1G APOSPC5438-500MG APOSPC5438-5G APOSPC5438-10G
2-(Trifluoromethyl)-DL-phenylglycine 97%
2-(Trifluoromethyl)-DL-phenylglycine
Formel: C₉H₈F₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD02662403
CAS-Nummer: 240490-00-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR