2-Chlor-6-fluorzimtsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlor-6-fluorcinnamsäure, 3-(2-Chlor-6-fluorphenyl)propensäure

PC5447-5G
APOSPC5447-5GEA 23.2 EUR
APOSPC5447-5G
2-Chlor-6-fluorzimtsäure 98%
2-Chlor-6-fluorzimtsäure
Formel: C₉H₆ClFO₂
Molecular Weight: 200,6 g/mol
Schmelzpunkt: 164…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051582
CAS-Nummer: 392-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR