5-[3-(Trifluoromethyl)phenyl]-2H-1,2,3,4-tetraazole 97%

Lieferant: Apollo Scientific
Achtung

PC5454-5G
APOSPC5454-5GEA 140 EUR
APOSPC5454-5G
5-[3-(Trifluoromethyl)phenyl]-2H-1,2,3,4-tetraazole 97%
5-[3-(Trifluoromethyl)phenyl]-2H-1,2,3,4-tetraazole
Formel: C₈H₅F₃N₄
Molecular Weight: 214.15 g/mol
Schmelzpunkt: 156…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068098
CAS-Nummer: 92712-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR