2'-(Difluoromethoxy)acetophenone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-(Difluoromethoxy)phenyl)ethanone

PC5464-10G PC5464-5G PC5464-1G PC5464-25G
APOSPC5464-10GEA 373 EUR
APOSPC5464-10G APOSPC5464-5G APOSPC5464-1G APOSPC5464-25G
2'-(Difluoromethoxy)acetophenone 98%
2'-(Difluoromethoxy)acetophenone
Formel: C₉H₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00236219
CAS-Nummer: 127842-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR