2-Methyl-6-(trifluoromethyl)nicotinoyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-6-(trifluoromethyl)pyridine-3-carbonyl chloride, 3-(Chlorocarbonyl)-2-methyl-6-(trifluoromethyl)pyridine
PC5468-5G
APOSPC5468-5GEA
219
EUR
APOSPC5468-5G
2-Methyl-6-(trifluoromethyl)nicotinoyl chloride
2-Methyl-6-(trifluoromethyl)nicotinoyl chloride
Formel:
C₈H₅ClF₃NO Molecular Weight: 223,58 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD02091398 CAS-Nummer: 261635-98-7 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...