2-(4-Fluorobenzoyl)benzoic acid

Lieferant: Apollo Scientific

PC5469-1G PC5469-250MG
APOSPC5469-1GEA 0 EUR
APOSPC5469-1G APOSPC5469-250MG
2-(4-Fluorobenzoyl)benzoic acid
2-(4-Fluorobenzoyl)benzoic acid
Formel: C₁₄H₉FO₃
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002473
CAS-Nummer: 7649-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR