2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole

Lieferant: Apollo Scientific

Synonyme: (2,2-Difluorobenzo[d][1,3]dioxol-5-yl)methanol, (2,2-Difluoro-1,3-benzodioxol-5-yl)methanol

PC5472-1G
APOSPC5472-1GEA 62.5 EUR
APOSPC5472-1G
2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole
2,2-Difluoro-5-(hydroxymethyl)-1,3-benzodioxole
Formel: C₈H₆F₂O₃
Molecular Weight: 188,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202699
CAS-Nummer: 72768-97-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR