3-Fluoro-4-(trifluoromethoxy)benzyl alcohol 98%

Lieferant: Apollo Scientific
Achtung

PC5474-1G
APOSPC5474-1GEA 0 EUR
APOSPC5474-1G
3-Fluoro-4-(trifluoromethoxy)benzyl alcohol 98%
3-Fluoro-4-(trifluoromethoxy)benzyl alcohol
Formel: C₈H₆F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660204
CAS-Nummer: 886498-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR