2-Methoxy-5-(trifluormethoxy)benzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5475-10G
PC5475-5G
PC5475-1G
PC5475-25G
APOSPC5475-10GEA
285
EUR
APOSPC5475-10G
APOSPC5475-5G
APOSPC5475-1G
APOSPC5475-25G
2-Methoxy-5-(trifluormethoxy)benzylalkohol 98%
2-Methoxy-5-(trifluormethoxy)benzylalkohol
Formel:
C₉H₉F₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115960 CAS-Nummer: 562840-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...