2-Methoxy-5-(trifluormethoxy)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC5475-10G PC5475-5G PC5475-1G PC5475-25G
APOSPC5475-10GEA 285 EUR
APOSPC5475-10G APOSPC5475-5G APOSPC5475-1G APOSPC5475-25G
2-Methoxy-5-(trifluormethoxy)benzylalkohol 98%
2-Methoxy-5-(trifluormethoxy)benzylalkohol
Formel: C₉H₉F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD04115960
CAS-Nummer: 562840-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR