2-Hydroxy-6-(trifluoromethyl)nicotinamide
Lieferant: Apollo Scientific
|
Achtung
|
PC5476-1G
APOSPC5476-1GEA
0
EUR
APOSPC5476-1G
2-Hydroxy-6-(trifluoromethyl)nicotinamide
2-Hydroxy-6-(trifluoromethyl)nicotinamide
Formel:
C₇H₅F₃N₂O₂ Schmelzpunkt: 220…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243729 CAS-Nummer: 116548-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...