5-Brom-2,2-difluor-1,3-benzodioxol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole
PC5489-100G
APOSPC5489-100GEA
63.8
EUR
APOSPC5489-100G
5-Brom-2,2-difluor-1,3-benzodioxol 98%
5-Brom-2,2-difluor-1,3-benzodioxol
Formel:
C₇H₃BrF₂O₂ Molecular Weight: 237 g/mol Siedepunkt: 78…79 °C/20mmHg Dichte: 1.74 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236212 CAS-Nummer: 33070-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...