5-Bromo-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile > 95%

Lieferant: Apollo Scientific
Achtung

PC5498-500MG
APOSPC5498-500MGEA 102 EUR
APOSPC5498-500MG
5-Bromo-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile > 95%
5-Bromo-2-hydroxy-6-methyl-4-(trifluoromethyl)nicotinonitrile
Formel: C₈H₄BrF₃N₂O
Molecular Weight: 281,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501053
CAS-Nummer: 1092352-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR