2-Methoxy-6-(trifluoromethyl)benzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC5505-10G PC5505-5G PC5505-1G
APOSPC5505-10GEA 0 EUR
APOSPC5505-10G APOSPC5505-5G APOSPC5505-1G
2-Methoxy-6-(trifluoromethyl)benzoic acid 98%
2-Methoxy-6-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09832319
CAS-Nummer: 119692-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR