3-Brom-5-fluor-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
PC5508-250MG
APOSPC5508-250MGEA
21.9
EUR
APOSPC5508-250MG
3-Brom-5-fluor-1H-indazol
3-Bromo-5-fluoro-1H-indazole
Formel:
C₇H₄BrFN₂ Molecular Weight: 215,0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781639 CAS-Nummer: 885519-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...