2-Bromotetrafluoropropionic acid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC5515-1G
APOSPC5515-1GEA
319
EUR
APOSPC5515-1G
2-Bromotetrafluoropropionic acid 97%
2-Bromotetrafluoropropionic acid
Formel:
C₃HBrF₄O₂ Siedepunkt: 60 °C/22mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD02183518 CAS-Nummer: 13859-31-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...