Novec HFE-71IPA Engineered Fluid (3M)

Lieferant: Apollo Scientific

PC5527-100G
APOSPC5527-100GEA 0 EUR
APOSPC5527-100G
Novec HFE-71IPA Engineered Fluid (3M)
Novec HFE-71IPA Engineered Fluid
Siedepunkt: 55 °C
Schmelzpunkt: –42 °C
Dichte: 1,42 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD23704319
CAS-Nummer: 185045-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR