3,3,4,4,4-Pentafluor-1-butanol 97%

Lieferant: Apollo Scientific
Achtung

PC5529-1G
APOSPC5529-1GEA 29 EUR
APOSPC5529-1G
3,3,4,4,4-Pentafluor-1-butanol 97%
3,3,4,4,4-Pentafluor-1-butanol
Formel: C₄H₅F₅O
Molecular Weight: 164,08 g/mol
Siedepunkt: 110…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03424479
CAS-Nummer: 54949-74-5
EINECS: 259-405-8
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR