6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine 97%

Lieferant: Apollo Scientific
Achtung

PC5532-1G
APOSPC5532-1GEA 48 EUR
APOSPC5532-1G
6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine 97%
6-Methyl-4-(trifluoromethyl)pyrid-2-yl hydrazine
Formel: C₇H₈F₃N₃
Molecular Weight: 191,16 g/mol
Schmelzpunkt: 82…83 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052901
CAS-Nummer: 22123-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR