4-Chloro-2-fluorophenylzinc iodide 0.5M in THF

Lieferant: Apollo Scientific

PC5552-50ML
APOSPC5552-50MLEA 952 EUR
APOSPC5552-50ML
4-Chloro-2-fluorophenylzinc iodide 0.5M in THF
4-Chloro-2-fluorophenylzinc iodide
Formel: C₆H₃ClFIZn
Molecular Weight: 321,853 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02260161
CAS-Nummer: 518989-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR