2,2-Difluoro-2-pyridin-2-ylacetamide ≥95%

Lieferant: Apollo Scientific
Achtung

PC5554-1G PC5554-250MG
APOSPC5554-1GEA 812 EUR
APOSPC5554-1G APOSPC5554-250MG
2,2-Difluoro-2-pyridin-2-ylacetamide ≥95%
2,2-Difluoro-2-pyridin-2-ylacetamide
Formel: C₇H₆F₂N₂O
Molecular Weight: 172.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858603
CAS-Nummer: 1064652-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR