2,2-Difluoro-2-pyridin-2-ylacetamide ≥95%
Lieferant: Apollo Scientific
|
Achtung
|
PC5554-1G
PC5554-250MG
APOSPC5554-1GEA
812
EUR
APOSPC5554-1G
APOSPC5554-250MG
2,2-Difluoro-2-pyridin-2-ylacetamide ≥95%
2,2-Difluoro-2-pyridin-2-ylacetamide
Formel:
C₇H₆F₂N₂O Molecular Weight: 172.132 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11858603 CAS-Nummer: 1064652-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...