Pentafluorobenzyl-p-toluenesulphonate 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC5557E-5G
APOSPC5557E-5GEA
0
EUR
APOSPC5557E-5G
Pentafluorobenzyl-p-toluenesulphonate 98%
Pentafluorobenzyl-p-toluenesulphonate
Formel:
C₁₄H₉F₅O₃S Schmelzpunkt: 78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06248628 CAS-Nummer: 32974-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...