1,1,3,3,3-Pentafluorpropylmethylether 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1,1,3,3,3-pentafluoropropyl ether

PC5571-1G
APOSPC5571-1GEA 23.2 EUR
APOSPC5571-1G
1,1,3,3,3-Pentafluorpropylmethylether 97%
1,1,3,3,3-Pentafluorpropylmethylether
Formel: C₄H₅F₅O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 886762-12-5
UN: 3271
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR