1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxamide 97%

Lieferant: Apollo Scientific
Achtung

PC5573-1G PC5573-500MG
APOSPC5573-1GEA 0 EUR
APOSPC5573-1G APOSPC5573-500MG
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxamide 97%
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxamide
Formel: C₁₁H₁₃F₃N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD03237056
CAS-Nummer: 465514-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR