5H-Perfluoro-3,4-bis(trifluoromethyl)hexa-2,4-diene 98%

Lieferant: Apollo Scientific

PC5579-1G
APOSPC5579-1GEA 379 EUR
APOSPC5579-1G
5H-Perfluoro-3,4-bis(trifluoromethyl)hexa-2,4-diene 98%
5H-Perfluoro-3,4-bis(trifluoromethyl)hexa-2,4-diene
Formel: C₈HF₁₃
Siedepunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06248650
CAS-Nummer: 104047-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR