5-(Difluoromethyl)nicotinaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC55850-1G
APOSPC55850-1GEA 1010 EUR
APOSPC55850-1G
5-(Difluoromethyl)nicotinaldehyde 95%
5-(Difluoromethyl)nicotinaldehyde
Formel: C₇H₅F₂NO
MDL: MFCD18256105
CAS-Nummer: 1211583-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR