Methyl 5-(difluoromethyl)nicotinate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-(difluoromethyl)pyridine-3-carboxylate

PC55854-1G
APOSPC55854-1GEA 1010 EUR
APOSPC55854-1G
Methyl 5-(difluoromethyl)nicotinate 95%
Methyl 5-(difluoromethyl)nicotinate
Formel: C₈H₇F₂NO₂
Molecular Weight: 187,14 g/mol
MDL: MFCD25476641
CAS-Nummer: 1550967-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR