5-(Difluoromethyl)pyridin-3-amine, 3-BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl N-[5-(difluoromethyl)pyridin-3-yl]carbamate

PC55856-1G
APOSPC55856-1GEA 1050 EUR
APOSPC55856-1G
5-(Difluoromethyl)pyridin-3-amine, 3-BOC protected 95%
5-(Difluoromethyl)pyridin-3-amine, 3-BOC protected
Formel: C₁₁H₁₄F₂N₂O₂
Molecular Weight: 244,24 g/mol
MDL: MFCD27981250
CAS-Nummer: 899899-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR