Z-1-Methoxy-3,3,3-trifluoropropene 92%
Lieferant: Apollo Scientific
|
Achtung
|
PC5593-5G
APOSPC5593-5GEA
176
EUR
APOSPC5593-5G
Z-1-Methoxy-3,3,3-trifluoropropene 92%
Z-1-Methoxy-3,3,3-trifluoropropene
Formel:
C₄H₅F₃O Molecular Weight: 126,08 g/mol Siedepunkt: 102…104 °C Dichte: 1,1567 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038317 CAS-Nummer: 26885-67-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...