Potassium (3,3,3-trifluoroprop-1-yl)trifluoroborate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Potassium trifluoro(3,3,3-trifluoroprop-1-yl)borate
PC56001-1G
APOSPC56001-1GEA
338
EUR
APOSPC56001-1G
Potassium (3,3,3-trifluoroprop-1-yl)trifluoroborate
Potassium (3,3,3-trifluoroprop-1-yl)trifluoroborate
Formel:
C₃H₄BF₆·K Molecular Weight: 203,964 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09993011 CAS-Nummer: 1430722-07-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...