5-Bromo-6-fluoro-N-methylisatoic anhydride

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-fluoro-1-methyl-2H-3,1-benzoxazine-2,4(1H)-dione

PC56003-1G PC56003-250MG
APOSPC56003-1GEA 905 EUR
APOSPC56003-1G APOSPC56003-250MG
5-Bromo-6-fluoro-N-methylisatoic anhydride
5-Bromo-6-fluoro-N-methylisatoic anhydride
Formel: C₉H₅BrFNO₃
Molecular Weight: 274.043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28897200
CAS-Nummer: 1980053-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR