1,1,3,3-Tetrafluoropropane (HFC-254fa) 97%

Lieferant: Apollo Scientific

PC5601-5G
APOSPC5601-5GEA 634 EUR
APOSPC5601-5G
1,1,3,3-Tetrafluoropropane (HFC-254fa) 97%
1,1,3,3-Tetrafluoropropane (HFC-254fa)
Formel: C₃H₄F₄
Molecular Weight: 116,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784205
CAS-Nummer: 66794-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR