3-Bromo-5-(trifluoromethyl)isoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-5-(trifluoromethyl)-1,2-oxazole

PC56013-100MG PC56013-250MG
APOSPC56013-100MGEA 377 EUR
APOSPC56013-100MG APOSPC56013-250MG
3-Bromo-5-(trifluoromethyl)isoxazole
3-Bromo-5-(trifluoromethyl)isoxazole
Formel: C₄HBrF₃NO
Molecular Weight: 215.956 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660139
CAS-Nummer: 933673-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR