Di-tert-butyl(methyl)phosphonium tetrafluoroborate 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC56018-5G
APOSPC56018-5GEA
155
EUR
APOSPC56018-5G
Di-tert-butyl(methyl)phosphonium tetrafluoroborate 95%
Di-tert-butyl(methyl)phosphonium tetrafluoroborate
Formel:
C₉H₂₂BF₄P Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840579 CAS-Nummer: 479094-62-7 EINECS: 808-098-4 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...