5-Bromo-3-hydroxy-2-nitrobenzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-nitro-3-(trifluoromethyl)phenol

PC56047-250MG
APOSPC56047-250MGEA 21.9 EUR
APOSPC56047-250MG
5-Bromo-3-hydroxy-2-nitrobenzotrifluoride
5-Bromo-3-hydroxy-2-nitrobenzotrifluoride
Formel: C₇H₃BrF₃NO₃
Molecular Weight: 286.003 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30746953
CAS-Nummer: 2092867-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR