3-(Trifluoromethylsulphonyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(Bromomethyl)-3-(trifluoromethylsulphonyl)benzene

PC56048-1G
APOSPC56048-1GEA 955 EUR
APOSPC56048-1G
3-(Trifluoromethylsulphonyl)benzyl bromide
3-(Trifluoromethylsulphonyl)benzyl bromide
Formel: C₈H₆BrF₃O₂S
Molecular Weight: 303.096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14706281
CAS-Nummer: 866208-07-3
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR