3-(Trifluoromethylsulphonyl)phenylacetonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
PC56067-1G
PC56067-250MG
APOSPC56067-1GEA
69.6
EUR
APOSPC56067-1G
APOSPC56067-250MG
3-(Trifluoromethylsulphonyl)phenylacetonitrile
3-(Trifluoromethylsulphonyl)phenylacetonitrile
Formel:
C₉H₆F₃NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD29065960 CAS-Nummer: 1301739-15-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...