1-(1,1-Difluoroethyl)-2-fluorobenzene 98%

Lieferant: Apollo Scientific
Achtung

PC56077-1G PC56077-250MG
APOSPC56077-1GEA 0 EUR
APOSPC56077-1G APOSPC56077-250MG
1-(1,1-Difluoroethyl)-2-fluorobenzene 98%
1-(1,1-Difluoroethyl)-2-fluorobenzene
Formel: C₈H₇F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11845814
CAS-Nummer: 1138445-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR