5-Methyl-2-(trifluoromethyl)-3-furoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Methyl-2-(trifluoromethyl)furan-3-carboxylic acid
PC5608-1G
APOSPC5608-1GEA
87
EUR
APOSPC5608-1G
5-Methyl-2-(trifluoromethyl)-3-furoic acid
5-Methyl-2-(trifluoromethyl)-3-furoic acid
Formel:
C₇H₅F₃O₃ Molecular Weight: 194,108 g/mol Schmelzpunkt: 124…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00275563 CAS-Nummer: 17515-74-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...