2-(Difluoromethyl)-6-fluorobenzonitrile

Lieferant: Apollo Scientific
Achtung

PC56085-1G PC56085-250MG
APOSPC56085-1GEA 0 EUR
APOSPC56085-1G APOSPC56085-250MG
2-(Difluoromethyl)-6-fluorobenzonitrile
2-(Difluoromethyl)-6-fluorobenzonitrile
Formel: C₈H₄F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD18395392
CAS-Nummer: 1261442-19-6
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR