5-Fluor-2-(trifluormethyl)benzonitril
Lieferant: Apollo Scientific
|
Gefahr
|
PC5609-1G
PC5609-500MG
PC5609-5G
PC5609-25G
APOSPC5609-1GEA
21.9
EUR
APOSPC5609-1G
APOSPC5609-500MG
APOSPC5609-5G
APOSPC5609-25G
5-Fluor-2-(trifluormethyl)benzonitril
5-Fluor-2-(trifluormethyl)benzonitril
Formel:
C₈H₃F₄N Molecular Weight: 189.11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236281 CAS-Nummer: 240800-45-7 UN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...