1-(3,3,3-Trifluoropropyl)piperazine

Lieferant: Apollo Scientific
Achtung

PC56127-1G
APOSPC56127-1GEA 815 EUR
APOSPC56127-1G
1-(3,3,3-Trifluoropropyl)piperazine
1-(3,3,3-Trifluoropropyl)piperazine
Formel: C₇H₁₃F₃N₂
Molecular Weight: 182,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16769787
CAS-Nummer: 399580-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR