2-Fluoro-4-(methylthio)pyridine

Lieferant: Apollo Scientific
Achtung

PC56138-1G PC56138-250MG
APOSPC56138-1GEA 433 EUR
APOSPC56138-1G APOSPC56138-250MG
2-Fluoro-4-(methylthio)pyridine
2-Fluoro-4-(methylthio)pyridine
Formel: C₆H₆FNS
Molecular Weight: 143.182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23713127
CAS-Nummer: 1428234-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR