2-Hydroxy-5-(pentafluorothio)benzoic acid 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Hydroxy-5-(pentafluorosulphanyl)benzoic acid
PC56143-100MG
APOSPC56143-100MGEA
511
EUR
APOSPC56143-100MG
2-Hydroxy-5-(pentafluorothio)benzoic acid 95+%
2-Hydroxy-5-(pentafluorothio)benzoic acid
Formel:
C₇H₅F₅O₃S Molecular Weight: 264,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31561289 CAS-Nummer: 1801700-81-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...